Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYB5D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254755.1 | NP_001241684.1 | 4 | 1183 | ||
NM_001254756.1 | NP_001241685.1 | 4 | 1186 | |||
NM_144611.3 | NP_653212.1 | 4 | 1878 | |||
XM_006721447.2 | XP_006721510.2 | 4 | 3015 | |||
XM_011523649.2 | XP_011521951.1 | 4 | 1266 | |||
XM_017024180.1 | XP_016879669.1 | 4 | 1269 | |||
XM_017024181.1 | XP_016879670.1 | 4 | 1158 | |||
XM_017024182.1 | XP_016879671.1 | 4 | 3320 | |||
GenBank mRNA | AK172844.1 | - | 4 | 1344 | ||
BC051697.1 | - | 4 | 1790 | 5190840 |