Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNRPD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278656.1 | NP_001265585.1 | 2 | 542 | ||
NM_004175.4 | NP_004166.1 | 2 | 556 | |||
NR_103819.1 | 2 | 556 | ||||
GenBank mRNA | AB451249.1 | - | 1 | 116 | ||
AB451373.1 | - | 1 | 116 | |||
AK296173.1 | - | 2 | 199 | |||
BC000457.2 | - | 2 | 212 | 2822042 | ||
BC003150.1 | - | 2 | 212 | 2822042 | ||
BC034447.1 | - | 2 | 239 | |||
BF344644.1 | - | 2 | 551 | |||
BM928709.1 | - | 2 | 504 | |||
BP202359.1 | - | 2 | 206 | |||
BY993733.1 | - | 2 | 228 | |||
CN286602.1 | - | 2 | 196 | |||
CR456583.1 | - | 2 | 232 | |||
U15009.1 | - | 2 | 203 |