Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEPSIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144679.2 | NP_653280.1 | 5 | 410 | ||
XM_005257066.2 | XP_005257123.1 | 6 | 1004 | |||
XM_005257067.2 | XP_005257124.1 | 5 | 410 | |||
XM_006721709.3 | XP_006721772.1 | 5 | 417 | |||
XM_006721712.2 | XP_006721775.1 | 5 | 848 | |||
XM_011524355.1 | XP_011522657.1 | 6 | 1010 | |||
XM_011524356.1 | XP_011522658.1 | 6 | 1010 | |||
XM_011524357.1 | XP_011522659.1 | 5 | 1038 | |||
XM_011524358.2 | XP_011522660.1 | 6 | 939 | |||
XM_017024202.1 | XP_016879691.1 | 6 | 1251 | |||
XM_017024203.1 | XP_016879692.1 | 7 | 898 | |||
XM_017024204.1 | XP_016879693.1 | 6 | 1451 | |||
GenBank mRNA | AK056090.1 | - | 5 | 374 | ||
AK127221.1 | - | 3 | 1563 | |||
AK128728.1 | - | 6 | 1723 | |||
BC064483.1 | - | 5 | 371 | 5430837 |