Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLFN13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144682.5 | NP_653283.3 | 3 | 1168 | ||
XM_005257922.2 | XP_005257979.1 | 3 | 1149 | |||
XM_017024232.1 | XP_016879721.1 | 2 | 2690 | |||
XM_017024233.1 | XP_016879722.1 | 3 | 2176 | |||
GenBank mRNA | AK126184.1 | - | 1 | 210 | ||
AK127728.1 | - | 1 | 1221 | |||
AL832726.1 | - | 3 | 1099 | |||
BC136622.1 | - | 3 | 1069 | 9020612 | ||
BC171771.1 | - | 3 | 1069 | 9054425 | ||
CR749630.1 | - | 3 | 1085 |