Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RILPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145058.2 | NP_659495.1 | 1 | 441 | ||
NR_130703.1 | 1 | 441 | ||||
XM_011538012.2 | XP_011536314.1 | 1 | 385 | |||
XM_017018959.1 | XP_016874448.1 | 1 | 385 | |||
GenBank mRNA | AB085763.1 | - | 1 | 204 | ||
AK056934.1 | - | 1 | 425 | |||
BC013042.2 | - | 1 | 214 | 3544761 | ||
BM556243.1 | - | 1 | 350 |