Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286256.1 | NP_001273185.1 | 11 | 1479 | ||
NM_001286257.1 | NP_001273186.1 | 9 | 1113 | |||
NM_145064.2 | NP_659501.1 | 12 | 1546 | |||
NR_104422.1 | 8 | 1046 | ||||
XM_011538126.2 | XP_011536428.1 | 12 | 1634 | |||
GenBank mRNA | AK057013.1 | - | 12 | 1539 | ||
AK311513.1 | - | 8 | 1039 | |||
BC008069.2 | - | 11 | 1459 | 2961089 |