Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256442.1 | NP_001243371.1 | 3 | 2394 | ||
NM_001256443.1 | NP_001243372.1 | 2 | 2821 | |||
NM_145239.2 | NP_660282.2 | 4 | 2232 | |||
XM_011545715.2 | XP_011544017.1 | 3 | 2323 | |||
XM_011545716.2 | XP_011544018.1 | 3 | 2339 | |||
XM_017022888.1 | XP_016878377.1 | 4 | 2161 | |||
GenBank mRNA | AK074572.1 | - | 4 | 2120 | ||
AK092265.1 | - | 2 | 2666 | |||
AK098379.1 | - | 6 | 1899 | |||
AK225785.1 | - | 4 | 2127 | |||
AL834185.1 | - | 4 | 2073 | |||
BC011405.1 | - | 4 | 2123 | 3868330 | ||
BC053594.1 | - | 4 | 2051 | 5729288 |