Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZG16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145252.2 | NP_660295.2 | 4 | 426 | ||
GenBank mRNA | AK311810.1 | - | 4 | 294 | ||
AY359021.1 | - | 4 | 368 | |||
BC009722.1 | - | 4 | 366 | 3879368 | ||
BM786200.1 | - | 2 | 135 | |||
BQ924772.1 | - | 4 | 429 | |||
EF067313.1 | - | 4 | 325 | |||
JN411265.1 | - | 4 | 367 |