Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZG16B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145252.2 | NP_660295.2 | 4 | 504 | ||
GenBank mRNA | AK311810.1 | - | 4 | 372 | ||
AY359021.1 | - | 4 | 446 | |||
BC009722.1 | - | 4 | 444 | 3879368 | ||
BM786200.1 | - | 2 | 213 | |||
BQ924772.1 | - | 4 | 507 | |||
EF067313.1 | - | 4 | 403 | |||
JN411265.1 | - | 4 | 445 |