Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM170A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304996.1 | NP_001291925.1 | 3 | 636 | ||
NM_001304997.1 | NP_001291926.1 | 3 | 620 | |||
NM_001304998.1 | NP_001291927.1 | 3 | 432 | |||
NM_145254.2 | NP_660297.1 | 3 | 705 | |||
XM_011522869.2 | XP_011521171.1 | 4 | 727 | |||
XM_017022939.1 | XP_016878428.1 | 3 | 962 | |||
XM_017022940.1 | XP_016878429.1 | 4 | 656 | |||
XM_017022941.1 | XP_016878430.1 | 3 | 521 | |||
XM_017022942.1 | XP_016878431.1 | 3 | 899 | |||
XM_017022943.1 | XP_016878432.1 | 3 | 568 | |||
GenBank mRNA | AK094930.1 | - | 3 | 620 | ||
AK298475.1 | - | 3 | 588 | |||
BC018082.1 | - | 3 | 654 | 4796761 | ||
BX647432.1 | - | 3 | 588 | |||
BX648429.1 | - | 3 | 589 | |||
BX648484.1 | - | 3 | 589 | |||
DA370427.1 | - | 3 | 432 |