Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA3F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318798.1 | NP_001305727.1 | 4, 5 | 369 | ||
NM_001318800.1 | NP_001305729.1 | 4, 5 | 653 | |||
NM_004186.4 | NP_004177.3 | 4, 5 | 537 | |||
XM_005265381.4 | XP_005265438.1 | 4, 5 | 667 | |||
XM_005265382.4 | XP_005265439.1 | 4, 5 | 470 | |||
XM_006713290.3 | XP_006713353.1 | 4, 5 | 470 | |||
XM_011533998.2 | XP_011532300.1 | 4, 5 | 469 | |||
XM_011534000.2 | XP_011532302.1 | 4, 5 | 470 | |||
GenBank mRNA | AB209259.1 | - | 4, 5 | 323 | ||
BC042914.1 | - | 4, 5 | 533 | 5477898 | ||
BF058597.1 | - | 4, 5 | 537 | |||
BQ949723.1 | - | 4, 5 | 478 | |||
U33920.1 | - | 3, 4 | 413 | |||
U38276.1 | - | 4, 5 | 650 |