Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319661.1 | NP_001306590.1 | 3 | 866 | ||
NM_001319662.1 | NP_001306591.1 | 3 | 887 | |||
NM_001319663.1 | NP_001306592.1 | 5 | 1032 | |||
NM_001319664.1 | NP_001306593.1 | 5 | 1050 | |||
NM_001319666.1 | NP_001306595.1 | 5 | 1053 | |||
NM_152261.3 | NP_689474.1 | 4 | 934 | |||
XM_017020198.1 | XP_016875687.1 | 4 | 1121 | |||
GenBank mRNA | AK001731.1 | - | 4 | 748 | ||
AK021783.1 | - | 2 | 549 | |||
AK022770.1 | - | 3 | 648 | |||
AK024021.1 | - | 4 | 761 | |||
BC020522.1 | - | 4 | 729 | 3916886 |