Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319661.1 | NP_001306590.1 | 3 | 1560 | ||
NM_001319662.1 | NP_001306591.1 | 3 | 1581 | |||
NM_001319663.1 | NP_001306592.1 | 5 | 1726 | |||
NM_001319664.1 | NP_001306593.1 | 5 | 1744 | |||
NM_001319666.1 | NP_001306595.1 | 5 | 1747 | |||
NM_152261.3 | NP_689474.1 | 4 | 1628 | |||
XM_017020198.1 | XP_016875687.1 | 4 | 1815 | |||
GenBank mRNA | AK001731.1 | - | 4 | 1442 | ||
AK021783.1 | - | 2 | 1243 | |||
AK022770.1 | - | 3 | 1342 | |||
AK024021.1 | - | 4 | 1455 | |||
BC020522.1 | - | 4 | 1423 | 3916886 |