Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143905.2 | NP_001137377.1 | 3 | 1295 | ||
NM_001194995.1 | NP_001181924.1 | 3 | 1588 | |||
NM_152269.4 | NP_689482.1 | 3 | 1379 | |||
XM_005253630.4 | XP_005253687.1 | 4 | 1399 | |||
XM_011538980.2 | XP_011537282.1 | 3 | 2157 | |||
GenBank mRNA | AF061733.1 | - | 4 | 1050 | ||
AK095982.1 | - | 3 | 1759 | |||
BC062329.1 | - | 3 | 1222 | 5484911 |