Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF641 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172681.1 | NP_001166152.1 | 6 | 707 | ||
NM_001172682.1 | NP_001166153.1 | 5 | 613 | |||
NM_152320.2 | NP_689533.2 | 7 | 880 | |||
XM_005268637.3 | XP_005268694.1 | 5 | 660 | |||
XM_005268638.3 | XP_005268695.1 | 6 | 900 | |||
XM_005268639.2 | XP_005268696.1 | 5 | 551 | |||
XM_011537897.1 | XP_011536199.1 | 6 | 880 | |||
XM_011537898.1 | XP_011536200.1 | 6 | 808 | |||
XM_017018797.1 | XP_016874286.1 | 5 | 704 | |||
XM_017018798.1 | XP_016874287.1 | 6 | 734 | |||
GenBank mRNA | AK055857.1 | - | 6 | 783 | ||
AK092740.1 | - | 6 | 1307 | |||
AK298064.1 | - | 5 | 605 | |||
AY842285.1 | - | 6 | 1334 | |||
BC018090.1 | - | 6 | 707 | 4791876 |