Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152345.4 | NP_689558.4 | 5 | 604 | ||
XM_005257920.4 | XP_005257977.1 | 5 | 537 | |||
XM_011524319.2 | XP_011522621.1 | 5 | 774 | |||
XM_011524320.2 | XP_011522622.1 | 5 | 536 | |||
XM_011524321.2 | XP_011522623.1 | 5 | 536 | |||
XM_017024174.1 | XP_016879663.1 | 5 | 471 | |||
XM_017024175.1 | XP_016879664.1 | 5 | 473 | |||
XM_017024176.1 | XP_016879665.1 | 5 | 403 | |||
GenBank mRNA | AK092673.1 | - | 5 | 421 | ||
AK097716.1 | - | 4 | 440 | |||
BC032554.1 | - | 5 | 651 | 5557374 | ||
BC062597.1 | - | 4 | 454 | |||
DA445860.1 | - | 5 | 471 |