Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163673.1 | NP_001157145.1 | 10 | 2032 | ||
NM_001163809.1 | NP_001157281.1 | 10 | 5717 | |||
NM_001163811.1 | NP_001157283.1 | 10 | 2168 | |||
NM_152348.3 | NP_689561.2 | 11 | 2669 | |||
XM_011523651.2 | XP_011521953.1 | 11 | 2654 | |||
XM_017024184.1 | XP_016879673.1 | 10 | 5643 | |||
GenBank mRNA | AK074111.1 | - | 9 | 3292 | ||
AK074356.1 | - | 5 | 1259 | |||
AK091136.1 | - | 11 | 2653 | |||
AK122972.1 | - | 10 | 2979 | |||
AK123896.1 | - | 10 | 2159 | |||
AK127946.1 | - | 10 | 2168 | |||
AK298567.1 | - | 10 | 2032 | |||
AK300986.1 | - | 8 | 1649 | |||
AL834379.1 | - | 10 | 2493 | |||
BC092513.1 | - | 10 | 2444 | 3092839 | ||
BC114519.1 | - | 9 | 1927 | 40036610 | ||
BC114568.1 | - | 10 | 1994 | 40036619 |