Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LRRC75A-AS1 SIRNA Assays ›
Gene Symbol:
LRRC75A-AS1
Gene Name:
LRRC75A antisense RNA 1
Gene Aliases:
C17orf45, C17orf76-AS1, FAM211A-AS1, NCRNA00188, TSAP19
Chromosome Location:
Chr.17: 16438987 - 16470648 on Build GRCh38
UniGene ID:
Hs.368934
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
141459
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LRRC75A-AS1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NR_027158.1
3
193
NR_027159.1
2
459
NR_027160.1
3
582
NR_027161.1
3
635
NR_027162.1
3
635
NR_027163.1
4
369
NR_027164.1
4
316
NR_027165.1
4
237
NR_027166.1
3
193
NR_027167.1
4
316
NR_027168.2
4
285
NR_027169.1
4
369
NR_027170.1
4
237
NR_027171.1
2
459
NR_027172.2
3
162
NR_027173.1
3
193
NR_027174.1
3
193
NR_027175.1
3
193
NR_027176.1
3
193
NR_027177.1
3
193
NR_027178.1
3
193
NR_027179.1
3
193
NR_027667.1
3
503
NR_045021.1
3
172
NR_045022.1
4
311
NR_045023.1
3
193
NR_045024.1
2
459
NR_045025.1
2
105
NR_045026.1
4
369
NR_045027.1
2
459
NR_045028.1
3
503
NR_045029.1
3
267
GenBank mRNA
AK098643.1
-
3
143
AK307982.1
-
3
585
AK308482.1
-
3
143
AK309961.1
-
4
267
AK311247.1
-
3
749
BC012396.1
-
2
113
BC019833.1
-
2
119
BC027986.1
-
3
453
4940349
BC040159.1
-
3
432
5539041
BI333700.1
-
3
264
BP353162.1
-
4
319
BP377665.1
-
2
394
BP870476.1
-
4
187
BQ070126.1
-
2
197
BQ954204.1
-
3
559
BU165462.1
-
2
386
BU506593.1
-
300
BU599615.1
-
4
323
BX538291.1
-
4
188
CB150672.1
-
4
266
CD050014.1
-
3
166
CF126952.1
-
2
375
CN297624.1
-
3
162
CR989654.1
-
3
246
DA131201.1
-
3
143
DA193585.1
-
3
328
DA383448.1
-
3
143
DA440445.1
-
4
262
DA485648.1
-
3
144
DA494627.1
-
2
410
DA769571.1
-
3
143
DA793334.1
-
2
59
DA802938.1
-
3
143
DA857697.1
-
3
143
DA867958.1
-
4
321
DB258221.1
-
2
410
DW409012.1
-
3
160
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline