Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC75A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027158.1 | 4 | 872 | |||
NR_027159.1 | 4 | 1195 | ||||
NR_027160.1 | 5 | 1318 | ||||
NR_027161.1 | 4 | 1314 | ||||
NR_027162.1 | 5 | 1371 | ||||
NR_027163.1 | 6 | 1105 | ||||
NR_027164.1 | 6 | 1052 | ||||
NR_027165.1 | 5 | 916 | ||||
NR_027166.1 | 5 | 929 | ||||
NR_027167.1 | 5 | 995 | ||||
NR_027168.2 | 6 | 1021 | ||||
NR_027169.1 | 5 | 1048 | ||||
NR_027170.1 | 6 | 973 | ||||
NR_027667.1 | 5 | 1239 | ||||
NR_045021.1 | 5 | 908 | ||||
NR_045022.1 | 6 | 1047 | ||||
NR_045027.1 | 3 | 1138 | ||||
NR_045028.1 | 4 | 1182 | ||||
NR_045029.1 | 5 | 1003 | ||||
GenBank mRNA | BC012396.1 | - | 3 | 791 | ||
BC019833.1 | - | 4 | 853 | |||
BC027986.1 | - | 5 | 1189 | 4940349 | ||
BC040159.1 | - | 5 | 1163 | 5539041 | ||
BX538291.1 | - | 6 | 923 |