Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMDHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152435.2 | NP_689648.2 | 7 | 1090 | ||
GenBank mRNA | AB075878.1 | - | 7 | 1045 | ||
AK054617.1 | - | 7 | 1090 | |||
AK122802.1 | - | 7 | 1057 | |||
AK290828.1 | - | 7 | 1045 | |||
AL833144.1 | - | 6 | 798 | |||
BC029146.1 | - | 7 | 1026 | 5183113 |