Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284417.1 | NP_001271346.1 | 3 | 1192 | ||
NM_001284418.1 | NP_001271347.1 | 3 | 1193 | |||
NM_001284419.1 | NP_001271348.1 | 5 | 1268 | |||
NM_001284420.1 | NP_001271349.1 | 2 | 902 | |||
NM_001284421.1 | NP_001271350.1 | 4 | 1218 | |||
NM_001284422.1 | NP_001271351.1 | 5 | 1318 | |||
NM_152449.3 | NP_689662.2 | 6 | 1383 | |||
XM_011521241.1 | XP_011519543.1 | 2 | 976 | |||
XM_011521244.1 | XP_011519546.1 | 3 | 1545 | |||
XM_017021926.1 | XP_016877415.1 | 4 | 1409 | |||
XM_017021927.1 | XP_016877416.1 | 3 | 1730 | |||
XM_017021928.1 | XP_016877417.1 | 2 | 1374 | |||
XM_017021929.1 | XP_016877418.1 | 4 | 1571 | |||
XM_017021930.1 | XP_016877419.1 | 4 | 1585 | |||
GenBank mRNA | AK057570.1 | - | 1 | 727 | ||
AK124852.1 | - | 4 | 1521 | |||
BC041097.1 | - | 6 | 1360 | 5240362 | ||
BC084545.1 | - | 3 | 1160 | 4590701 |