Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284417.1 | NP_001271346.1 | 3 | 2370 | ||
NM_001284418.1 | NP_001271347.1 | 3 | 2371 | |||
NM_001284419.1 | NP_001271348.1 | 5 | 2446 | |||
NM_001284420.1 | NP_001271349.1 | 2 | 2080 | |||
NM_001284421.1 | NP_001271350.1 | 4 | 2396 | |||
NM_001284422.1 | NP_001271351.1 | 5 | 2496 | |||
NM_152449.3 | NP_689662.2 | 6 | 2561 | |||
XM_011521241.1 | XP_011519543.1 | 2 | 2154 | |||
XM_011521244.1 | XP_011519546.1 | 3 | 2723 | |||
XM_017021926.1 | XP_016877415.1 | 4 | 2587 | |||
XM_017021927.1 | XP_016877416.1 | 3 | 2908 | |||
XM_017021928.1 | XP_016877417.1 | 2 | 2552 | |||
XM_017021929.1 | XP_016877418.1 | 4 | 2749 | |||
XM_017021930.1 | XP_016877419.1 | 4 | 2763 | |||
GenBank mRNA | AK057570.1 | - | 1 | 1905 | ||
BC041097.1 | - | 6 | 2538 | 5240362 | ||
BC084545.1 | - | 3 | 2338 | 4590701 |