Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM81A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152450.2 | NP_689663.2 | 5 | 642 | ||
XM_005254166.2 | XP_005254223.1 | 5 | 555 | |||
XM_006720398.3 | XP_006720461.1 | 7 | 824 | |||
XM_006720399.2 | XP_006720462.1 | 5 | 615 | |||
XM_011521247.2 | XP_011519549.1 | 7 | 788 | |||
XM_011521248.2 | XP_011519550.1 | 6 | 669 | |||
XM_011521249.2 | XP_011519551.1 | 6 | 779 | |||
XM_011521250.2 | XP_011519552.1 | 6 | 632 | |||
XM_017021931.1 | XP_016877420.1 | 6 | 1080 | |||
XM_017021932.1 | XP_016877421.1 | 5 | 670 | |||
GenBank mRNA | AK307975.1 | - | 5 | 585 | ||
BC022537.1 | - | 5 | 642 | 4817144 |