Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM81A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152450.2 | NP_689663.2 | 5 | 669 | ||
XM_005254166.2 | XP_005254223.1 | 5 | 582 | |||
XM_006720398.3 | XP_006720461.1 | 7 | 851 | |||
XM_006720399.2 | XP_006720462.1 | 5 | 642 | |||
XM_011521247.2 | XP_011519549.1 | 7 | 815 | |||
XM_011521248.2 | XP_011519550.1 | 6 | 696 | |||
XM_011521249.2 | XP_011519551.1 | 6 | 806 | |||
XM_011521250.2 | XP_011519552.1 | 6 | 659 | |||
XM_017021931.1 | XP_016877420.1 | 6 | 1107 | |||
XM_017021932.1 | XP_016877421.1 | 5 | 697 | |||
GenBank mRNA | AK307975.1 | - | 5 | 612 | ||
BC022537.1 | - | 5 | 669 | 4817144 |