Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152453.3 | NP_689666.2 | 3 | 263 | ||
XM_005254168.3 | XP_005254225.1 | 3 | 244 | |||
XM_011521262.2 | XP_011519564.1 | 3 | 244 | |||
XM_011521263.1 | XP_011519565.1 | 3 | 244 | |||
XM_011521264.1 | XP_011519566.1 | 3 | 244 | |||
GenBank mRNA | AK093126.1 | - | 3 | 244 | ||
AK310517.1 | - | 3 | 249 | |||
BC029221.2 | - | 2 | 176 | 5167874 | ||
BI561792.1 | - | 3 | 267 | |||
BI826726.1 | - | 3 | 227 |