Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152455.3 | NP_689668.3 | 5 | 1964 | ||
XM_006720401.3 | XP_006720464.1 | 6 | 2724 | |||
XM_011521266.2 | XP_011519568.1 | 6 | 2404 | |||
GenBank mRNA | AF525399.1 | - | 5 | 1814 | ||
AK093186.1 | - | 4 | 1510 | |||
AK093377.1 | - | 3 | 999 | |||
AK122788.1 | - | 5 | 1952 | |||
BC017179.2 | - | 3 | 830 | |||
BC040682.1 | - | 3 | 830 | |||
BC043612.1 | - | 5 | 2037 | |||
BC109270.1 | - | 5 | 1603 | 40026738 | ||
BC109271.1 | - | 3 | 647 | 40026744 |