Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C16orf89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098514.2 | NP_001091984.2 | 2 | 578 | ||
NM_152459.4 | NP_689672.4 | 2 | 578 | |||
XM_005255143.3 | XP_005255200.2 | 2 | 404 | |||
XM_011522392.2 | XP_011520694.1 | 1 | 70 | |||
XM_017022972.1 | XP_016878461.1 | 2 | 404 | |||
XM_017022973.1 | XP_016878462.1 | 2 | 404 | |||
XM_017022974.1 | XP_016878463.1 | 2 | 404 | |||
GenBank mRNA | AK075242.1 | - | 2 | 404 | ||
AK300708.1 | - | 2 | 352 | |||
AY358483.1 | - | 2 | 543 | |||
BC033681.1 | - | 2 | 473 | 5190553 |