Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM199 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152464.2 | NP_689677.1 | 3 | 437 | ||
GenBank mRNA | AK223584.1 | - | 3 | 371 | ||
AK291475.1 | - | 3 | 360 | |||
AK311363.1 | - | 319 | ||||
AY074907.1 | - | 3 | 347 | |||
BC033113.1 | - | 3 | 354 | 5429060 | ||
DC327155.1 | - | 3 | 437 |