Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286441.1 | NP_001273370.1 | 5 | 646 | ||
NM_152596.3 | NP_689809.2 | 3 | 391 | |||
XM_006720415.3 | XP_006720478.1 | 4 | 563 | |||
XM_011521298.2 | XP_011519600.1 | 6 | 876 | |||
XM_011521299.2 | XP_011519601.1 | 6 | 607 | |||
XM_011521300.2 | XP_011519602.1 | 3 | 135 | |||
XM_011521301.2 | XP_011519603.1 | 5 | 432 | |||
XM_017021970.1 | XP_016877459.1 | 3 | 135 | |||
XM_017021971.1 | XP_016877460.1 | 3 | 133 | |||
GenBank mRNA | AK292524.1 | - | 3 | 297 | ||
AK302864.1 | - | 5 | 580 | |||
BC030628.2 | - | 3 | 331 | 4827307 |