Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARCH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100875.2 | NP_001094345.1 | 5 | 680 | ||
NM_001288779.1 | NP_001275708.1 | 6 | 875 | |||
NM_001288780.1 | NP_001275709.1 | 6 | 906 | |||
NM_152598.3 | NP_689811.2 | 5 | 761 | |||
XM_005257095.2 | XP_005257152.2 | 6 | 837 | |||
XM_005257096.2 | XP_005257153.2 | 6 | 782 | |||
XM_005257097.3 | XP_005257154.2 | 7 | 929 | |||
XM_005257098.2 | XP_005257155.2 | 6 | 834 | |||
XM_005257100.2 | XP_005257157.2 | 5 | 723 | |||
XM_005257102.2 | XP_005257159.2 | 5 | 720 | |||
XM_005257103.1 | XP_005257160.1 | 5 | 720 | |||
XM_005257106.4 | XP_005257163.2 | 5 | 488 | |||
XM_011524435.1 | XP_011522737.1 | 6 | 834 | |||
XM_011524436.1 | XP_011522738.1 | 5 | 668 | |||
XM_011524437.1 | XP_011522739.1 | 6 | 812 | |||
GenBank mRNA | AK093076.1 | - | 5 | 714 | ||
AK122947.1 | - | 6 | 815 | |||
BC035021.1 | - | 5 | 684 | 4824637 | ||
BX648705.1 | - | 6 | 838 |