Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152638.3 | NP_689851.1 | 1 | 1318 | ||
GenBank mRNA | BC024183.2 | - | 1 | 1316 | 4822259 | |
BC026075.2 | - | 1 | 1316 | 4828455 | ||
BC027600.2 | - | 1 | 1318 | 4828721 | ||
BC034491.2 | - | 1 | 1316 | 4822257 |