Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152638.3 | NP_689851.1 | 1 | 2634 | ||
NR_130711.1 | 2 | 402 | ||||
GenBank mRNA | BC027600.2 | - | 1 | 2634 | 4828721 | |
BX114695.1 | - | 2 | 402 |