Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM227B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152647.2 | NP_689860.2 | 4 | 593 | ||
XM_005254213.3 | XP_005254270.1 | 4 | 670 | |||
XM_005254214.3 | XP_005254271.1 | 4 | 603 | |||
XM_005254215.3 | XP_005254272.1 | 4 | 670 | |||
XM_005254216.3 | XP_005254273.1 | 4 | 670 | |||
XM_005254219.3 | XP_005254276.1 | 4 | 670 | |||
XM_005254220.3 | XP_005254277.1 | 4 | 670 | |||
XM_006720423.2 | XP_006720486.1 | 5 | 635 | |||
XM_006720424.2 | XP_006720487.1 | 4 | 670 | |||
XM_006720426.2 | XP_006720489.1 | 4 | 670 | |||
XM_006720427.3 | XP_006720490.1 | 5 | 641 | |||
XM_011521319.2 | XP_011519621.1 | 4 | 2553 | |||
XM_011521321.2 | XP_011519623.1 | 4 | 670 | |||
XM_011521324.1 | XP_011519626.1 | 4 | 670 | |||
XM_011521325.2 | XP_011519627.1 | 4 | 670 | |||
XM_011521326.1 | XP_011519628.1 | 4 | 670 | |||
XM_017021990.1 | XP_016877479.1 | 4 | 670 | |||
XM_017021991.1 | XP_016877480.1 | 5 | 710 | |||
XM_017021993.1 | XP_016877482.1 | 4 | 670 | |||
XM_017021994.1 | XP_016877483.1 | 4 | 670 | |||
XM_017021995.1 | XP_016877484.1 | 4 | 670 | |||
GenBank mRNA | AK057362.1 | - | 4 | 591 | ||
BC048125.1 | - | 4 | 595 | 5272014 | ||
BC048139.1 | - | 4 | 595 |