Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142497.2 | NP_001135969.1 | 6 | 1641 | ||
NM_152649.3 | NP_689862.1 | 11 | 2265 | |||
XM_005255834.3 | XP_005255891.1 | 12 | 2164 | |||
GenBank mRNA | AK091708.1 | - | 11 | 2006 | ||
AK093013.1 | - | 3 | 2070 | |||
AK122998.1 | - | 3 | 3386 | |||
AL833362.1 | - | 10 | 2591 | |||
BC028141.1 | - | 6 | 1283 | 5214733 |