Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC47A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099646.1 | NP_001093116.1 | 9 | 968 | ||
NM_001256663.1 | NP_001243592.1 | 9 | 968 | |||
NM_152908.3 | NP_690872.2 | 9 | 1076 | |||
NR_135624.1 | 9 | 1539 | ||||
NR_135625.1 | 9 | 1122 | ||||
XM_017024221.1 | XP_016879710.1 | 9 | 1010 | |||
XM_017024222.1 | XP_016879711.1 | 9 | 1510 | |||
XM_017024223.1 | XP_016879712.1 | 9 | 1010 | |||
XM_017024224.1 | XP_016879713.1 | 9 | 1010 | |||
XM_017024225.1 | XP_016879714.1 | 9 | 1010 | |||
XM_017024226.1 | XP_016879715.1 | 9 | 1010 | |||
GenBank mRNA | AB250364.1 | - | 9 | 811 | ||
AB250701.1 | - | 9 | 965 | |||
AK055758.1 | - | 9 | 849 | |||
AK096080.1 | - | 8 | 1997 | |||
AK307647.1 | - | 9 | 968 | |||
AK310218.1 | - | 7 | 682 | |||
BC035288.1 | - | 9 | 836 | 5189385 | ||
BC050578.1 | - | 9 | 952 | 5759988 | ||
BG432484.1 | - | 1 | 53 | |||
BM924669.1 | - | 804 | ||||
BX648861.1 | - | 9 | 1539 |