Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQCK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305121.1 | NP_001292050.1 | 9 | 1983 | ||
NM_001305122.1 | NP_001292051.1 | 9 | 1963 | |||
NM_153208.2 | NP_694940.1 | 10 | 2048 | |||
NR_130967.1 | 8 | 1917 | ||||
NR_130968.1 | 9 | 1950 | ||||
XM_011545736.2 | XP_011544038.1 | 9 | 1383 | |||
XM_017022933.1 | XP_016878422.1 | 9 | 2094 | |||
GenBank mRNA | AF520569.1 | - | 8 | 1917 | ||
AF520570.1 | - | 10 | 2048 | |||
AK093894.1 | - | 7 | 1188 | |||
BC010515.1 | - | 4 | 859 | |||
BC034823.1 | - | 9 | 1379 | 5165639 |