Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286638.1 | NP_001273567.1 | 11 | 1393 | ||
NM_153251.3 | NP_694983.2 | 12 | 1416 | |||
NR_104486.1 | 15 | 1661 | ||||
NR_104487.1 | 13 | 1465 | ||||
XM_006719795.1 | XP_006719858.1 | 13 | 1437 | |||
XM_006719796.1 | XP_006719859.1 | 13 | 1434 | |||
XM_006719797.1 | XP_006719860.1 | 12 | 1359 | |||
XM_011535030.2 | XP_011533332.1 | 7 | 916 | |||
XM_017020525.1 | XP_016876014.1 | 11 | 1130 | |||
XM_017020526.1 | XP_016876015.1 | 14 | 1203 | |||
XM_017020527.1 | XP_016876016.1 | 12 | 1335 | |||
GenBank mRNA | AK295804.1 | - | 13 | 1370 | ||
AL833626.1 | - | 3 | 261 | |||
BC034944.1 | - | 15 | 1804 | |||
BC050367.1 | - | 12 | 1331 | 5591379 | ||
BC067898.1 | - | 15 | 1660 |