Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153260.2 | NP_694992.2 | 3 | 252 | ||
XM_011521423.2 | XP_011519725.1 | 2 | 140 | |||
XM_011521424.2 | XP_011519726.1 | 2 | 140 | |||
GenBank mRNA | AK094131.1 | - | 3 | 246 | ||
AK094891.1 | - | 2 | 486 | |||
AK309861.1 | - | 3 | 241 | |||
BC058935.1 | - | 3 | 208 | 5578024 | ||
BX538324.1 | - | 2 | 398 | |||
CR749223.1 | - | 3 | 221 | |||
DA749946.1 | - | 3 | 252 |