Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153356.2 | NP_699187.1 | 2 | 205 | ||
XM_006720409.3 | XP_006720472.1 | 2 | 235 | |||
XM_011521281.2 | XP_011519583.1 | 2 | 234 | |||
XM_011521282.2 | XP_011519584.1 | 2 | 234 | |||
XM_011521283.2 | XP_011519585.1 | 2 | 234 | |||
GenBank mRNA | BC033516.1 | - | 2 | 164 | 5170475 |