Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286434.1 | NP_001273363.1 | 3 | 309 | ||
NM_153356.2 | NP_699187.1 | 4 | 417 | |||
XM_006720409.3 | XP_006720472.1 | 4 | 447 | |||
XM_006720410.3 | XP_006720473.1 | 3 | 339 | |||
XM_006720411.3 | XP_006720474.1 | 2 | 235 | |||
XM_011521281.2 | XP_011519583.1 | 4 | 446 | |||
XM_011521282.2 | XP_011519584.1 | 4 | 446 | |||
XM_011521283.2 | XP_011519585.1 | 4 | 446 | |||
XM_011521284.2 | XP_011519586.1 | 3 | 226 | |||
GenBank mRNA | AB449907.1 | - | 3 | 185 | ||
BC033516.1 | - | 4 | 376 | 5170475 | ||
HY018107.1 | - | 2 | 206 |