Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286434.1 | NP_001273363.1 | 10 | 1104 | ||
NM_153356.2 | NP_699187.1 | 11 | 1212 | |||
GenBank mRNA | BC033516.1 | - | 11 | 1171 | 5170475 |