Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143917.1 | NP_001137389.1 | 3 | 792 | ||
NM_153374.2 | NP_699205.1 | 3 | 1383 | |||
XM_017022057.1 | XP_016877546.1 | 3 | 775 | |||
GenBank mRNA | AK129759.1 | - | 3 | 862 | ||
AY125955.1 | - | 3 | 1372 | |||
BC022075.1 | - | 3 | 922 | 4799858 | ||
BC033515.1 | - | 3 | 1380 | 5175226 | ||
CR936638.1 | - | 2 | 422 |