Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LPCAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153613.2 | NP_705841.2 | 2 | 339 | ||
XM_006720454.2 | XP_006720517.1 | 2 | 341 | |||
XM_011521422.1 | XP_011519724.1 | 2 | 321 | |||
GenBank mRNA | AF529233.1 | - | 2 | 190 | ||
AF542964.1 | - | 2 | 423 | |||
AK001119.1 | - | 1 | 385 | |||
AK290273.1 | - | 2 | 310 | |||
AK308910.1 | - | 2 | 321 | |||
AL832237.1 | - | 2 | 336 | |||
AY734233.1 | - | 2 | 359 | |||
BC048430.1 | - | 2 | 341 | |||
BC092463.1 | - | 2 | 336 | 30520144 | ||
BC144236.1 | - | 2 | 267 |