Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153634.2 | NP_705898.1 | 15 | 1213 | ||
XM_011537951.2 | XP_011536253.1 | 15 | 4188 | |||
XM_017018851.1 | XP_016874340.1 | 16 | 2186 | |||
XM_017018852.1 | XP_016874341.1 | 15 | 1159 | |||
GenBank mRNA | AK074223.1 | - | 1 | 75 | ||
AK094998.1 | - | 3 | 787 | |||
AK098593.1 | - | 15 | 1144 | |||
AK307908.1 | - | 15 | 1184 | |||
AY177785.1 | - | 15 | 1129 | |||
BC046366.1 | - | 3 | 530 | |||
BC110577.1 | - | 15 | 1204 | 40007743 | ||
BC110578.1 | - | 15 | 1204 | 40007744 | ||
BX538055.1 | - | 15 | 1227 |