Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144508.4 | NP_653091.3 | 10 | 907 | ||
NM_170589.4 | NP_733468.3 | 11 | 985 | |||
XM_017022432.1 | XP_016877921.1 | 9 | 1110 | |||
GenBank mRNA | AB022190.1 | - | 10 | 908 | ||
AF173994.1 | - | 11 | 1214 | |||
AF248041.1 | - | 10 | 891 | |||
AF461041.1 | - | 10 | 891 | |||
AK307605.1 | - | 10 | 899 | |||
AK307996.1 | - | 10 | 897 | |||
BC029373.1 | - | 10 | 884 |