Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18orf54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288980.1 | NP_001275909.1 | 9 | 4170 | ||
NM_001288981.1 | NP_001275910.1 | 9 | 4002 | |||
NM_001288982.1 | NP_001275911.1 | 7 | 3373 | |||
NM_173529.5 | NP_775800.3 | 8 | 3528 | |||
NR_110234.1 | 6 | 2854 | ||||
XM_005258201.1 | XP_005258258.1 | 8 | 4871 | |||
XM_005258203.1 | XP_005258260.1 | 8 | 4331 | |||
XM_017025567.1 | XP_016881056.1 | 8 | 4554 | |||
GenBank mRNA | AB267906.1 | - | 8 | 4862 | ||
AB330143.1 | - | 8 | 4379 | |||
AB330144.1 | - | 8 | 4322 | |||
AB353076.1 | - | 9 | 4165 | |||
AB353077.1 | - | 9 | 3997 | |||
BC043651.2 | - | 1 | 351 | |||
BX640877.1 | - | 8 | 3497 |