Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173591.3 | NP_775862.3 | 50 | 6122 | ||
XM_005268802.3 | XP_005268859.1 | 54 | 6923 | |||
XM_011538191.1 | XP_011536493.1 | 51 | 6199 | |||
XM_011538192.2 | XP_011536494.1 | 48 | 6140 | |||
XM_011538193.1 | XP_011536495.1 | 48 | 6059 | |||
GenBank mRNA | AK096852.1 | - | 4 | 400 | ||
BC101016.2 | - | 4 | 271 | |||
BC101017.2 | - | 4 | 271 | |||
BC101018.1 | - | 4 | 271 |