Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031748.3 | NP_001026918.2 | 8 | 1033 | ||
NM_001319247.1 | NP_001306176.1 | 8 | 1033 | |||
NR_135051.1 | 7 | 894 | ||||
XM_005268806.3 | XP_005268863.1 | 7 | 843 | |||
XM_011538231.2 | XP_011536533.1 | 7 | 1070 | |||
GenBank mRNA | AK097445.1 | - | 7 | 843 | ||
BC038754.1 | - | 8 | 1030 | 5269851 | ||
BC048120.1 | - | 8 | 1017 | 5266211 | ||
EU668334.1 | - | 7 | 843 | |||
GQ472216.1 | - | 7 | 843 |