Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSUH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011533774.1 | XP_011532076.1 | 1 | 76 | ||
XM_017006511.1 | XP_016862000.1 | 1 | 76 | |||
XM_017006512.1 | XP_016862001.1 | 1 | 75 | |||
XM_017006513.1 | XP_016862002.1 | 1 | 67 | |||
XM_017006514.1 | XP_016862003.1 | 1 | 76 | |||
XM_017006515.1 | XP_016862004.1 | 1 | 830 | |||
XM_017006518.1 | XP_016862007.1 | 1 | 76 | |||
XM_017006521.1 | XP_016862010.1 | 1 | 76 | |||
XM_017006522.1 | XP_016862011.1 | 1 | 76 | |||
XM_017006523.1 | XP_016862012.1 | 1 | 77 | |||
XM_017006524.1 | XP_016862013.1 | 1 | 76 | |||
XM_017006525.1 | XP_016862014.1 | 1 | 76 | |||
XM_017006527.1 | XP_016862016.1 | 1 | 76 | |||
XM_017006528.1 | XP_016862017.1 | 1 | 61 | |||
XM_017006533.1 | XP_016862022.1 | 1 | 76 | |||
XM_017006534.1 | XP_016862023.1 | 1 | 76 | |||
XM_017006538.1 | XP_016862027.1 | 1 | 76 | |||
GenBank mRNA | AK093066.1 | - | 5 | 958 | ||
AK122938.1 | - | 1 | 190 |
Gene Symbol: LINC01198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120426.1 | 1 | 155 | |||
GenBank mRNA | CN278417.1 | - | 1 | 155 | ||
HY006332.1 | - | 2 | 163 |
Gene Symbol: LOC101929998 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK127966.1 | - | 1 | 183 |
Gene Symbol: LOC102723968 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130763.1 | 2 | 900 | |||
GenBank mRNA | AK098560.1 | - | 2 | 900 | ||
AK127969.1 | - | 2 | 216 |
Gene Symbol: LOC107984014 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017012898.1 | XP_016868387.1 | 3 | 357 |
Gene Symbol: LOC440896 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015361.1 | 1 | 206 | |||
GenBank mRNA | AK055091.1 | - | 1 | 209 | ||
AK127288.1 | - | 1 | 1322 | |||
BC032955.1 | - | 4 | 865 |
Gene Symbol: OR7E156P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002171.2 | 1 | 902 | |||
GenBank mRNA | AK097469.1 | - | 1 | 902 | ||
AK097490.1 | - | 1 | 209 | |||
AK097573.1 | - | 1 | 180 |
Gene Symbol: OVOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX647938.1 | - | 7 | 4461 |
Gene Symbol: TPT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024458.1 | 6 | 613 | |||
GenBank mRNA | AF318337.1 | - | 6 | 613 | ||
AK124332.1 | - | 4 | 528 |