Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TVP23C-CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204478.1 | NP_001191407.1 | 7 | 2330 | ||
NR_037924.1 | 6 | 2185 | ||||
GenBank mRNA | AB074181.1 | - | 7 | 2260 |
Gene Symbol: CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204477.1 | NP_001191406.1 | 4 | 1919 | ||
GenBank mRNA | AK093993.1 | - | 2 | 1663 | ||
AK172733.1 | - | 1 | 1056 |